Learn More
Triforest Enterprises 1000mL Round Autoclaveable Polycarbonate Media Bottle
Catalog No. NC2401712
Encompass_Preferred
TriForest PC labware are ideal for steam sterilization of all liquids and buffers. The bottles withstand repeated autoclaving, and are optimal for preparation of terminally sterile fluids. Provided non sterile with blue autoclavable caps.
Catalog No. NC2401712 Supplier Triforest Enterprises Supplier No. RPC1000
TriForest PC labware are ideal for steam sterilization of all liquids and buffers. The bottles withstand repeated autoclaving, and are optimal for preparation of terminally sterile fluids. Provided non sterile with blue autoclavable caps.