Learn More
Ace Glass, Inc. Fabric heating mantle that covers top and bottom of 1, 2 or 3 neck 1000mL spherical flask. Lower half wattage is 380w at 115 volts. Upper half wattage is 140w at 115 volts, CSA rated.
SDP
Catalog No. 50977259
Encompass
Catalog No. 50-977-259 Supplier Ace Glass, Inc. Supplier No. 1203119