Learn More
NEST SCIENTIFIC INC 1000mL PET Square Media Bottles, Sterile, 12/cs
Catalog No. NC2887691
Encompass_Preferred
Nest Scientific square media bottles are molded from USP grade VI virgin PET. Packed in double layer, non-shedding bags for use in cleanroom production environments. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. Extensive validation dossiers available upon request.
Catalog No. NC2887691 Supplier NEST SCIENTIFIC INC Supplier No. 334001
Nest Scientific square media bottles are molded from USP grade VI virgin PET. Packed in double layer, non-shedding bags for use in cleanroom production environments. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. Extensive validation dossiers available upon request.