Learn More
Bellco Glass, Inc. 1000mL Square shape PETG Media Bottles, Sterile, 4/pk, 12/cs
SDP
Catalog No. NC3865169
Encompass_Preferred
1000mL PETG Square Media Bottless, Sterile, 4/pk, 12/cs. It is mainly used for storing and sampling active pharmaceutical ingredients, bulk intermediates, and also for preparing, storing buffers, culture solutions or long-term storage of pH sensitive liquids. Tips: Can't autoclave. It can be used to storage reagents but cannot be used as heating vessels. PETG Bottles are Suitable for handling and reagents and medium storing critical(protein/ vaccine etc.PET Bottles are used for storage of culture medium, serum, buffer and long-term storage of PH-sensitive liquid. Excellent physical and mechanical properties over a wide temperature range.Temperature Range: -80 oC to 50 oC.
Catalog No. NC3865169 Supplier Bellco Glass, Inc. Supplier No. 451400434/SQ1026866
1000mL PETG Square Media Bottless, Sterile, 4/pk, 12/cs. It is mainly used for storing and sampling active pharmaceutical ingredients, bulk intermediates, and also for preparing, storing buffers, culture solutions or long-term storage of pH sensitive liquids. Tips: Can't autoclave. It can be used to storage reagents but cannot be used as heating vessels. PETG Bottles are Suitable for handling and reagents and medium storing critical(protein/ vaccine etc.PET Bottles are used for storage of culture medium, serum, buffer and long-term storage of PH-sensitive liquid. Excellent physical and mechanical properties over a wide temperature range.Temperature Range: -80 oC to 50 oC.