Learn More
Chemglass Life Sciences 1000mL Storage Flask, Cajon, Airfree, Schlenk, 0-8mm Valve, 220mm OAH
SDP
Catalog No. AF009405
Encompass_Preferred
1000mL Storage Flask, Cajon, Airfree, Schlenk, 0-8mm Valve, 220mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.
Catalog No. AF009405 Supplier Chemglass Life Sciences Supplier No. AF009405
1000mL Storage Flask, Cajon, Airfree, Schlenk, 0-8mm Valve, 220mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.