Learn More
S2 Media 1L Tryptic Soy Broth (TSB) with 5% Sheep Blood, 1L Media Bottle, 1/each
Catalog No. 502995951
Encompass_Preferred
Tryptic Soy Broth (TSB) supplemented with 5% Sheep Blood is a non-selective and non-differential medium utilized for the cultivation of fastidious microorganisms. TSB is recommended for use as the inoculum broth for disc diffusion and agar dilution antimicrobial susceptibility testing by the CLSI. Supplemented product complies with ATCC Medium 260. Reference the appropriate standard method for additional details on test instructions. • Product base meets AOAC, BAM, CLSI, EPA, ISO, NF, SM-Dairy, SMWW, USDA, and Harmonized USP/EP/JP reference methods and performance criteria.• TSB with Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5951 Supplier S2 Media Supplier No. 5184
Tryptic Soy Broth (TSB) supplemented with 5% Sheep Blood is a non-selective and non-differential medium utilized for the cultivation of fastidious microorganisms. TSB is recommended for use as the inoculum broth for disc diffusion and agar dilution antimicrobial susceptibility testing by the CLSI. Supplemented product complies with ATCC Medium 260. Reference the appropriate standard method for additional details on test instructions. • Product base meets AOAC, BAM, CLSI, EPA, ISO, NF, SM-Dairy, SMWW, USDA, and Harmonized USP/EP/JP reference methods and performance criteria.• TSB with Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.