Learn More
Ace Glass, Inc. 1000mL vacuum trap 125 x 315mm w/#15 thread
SDP
Catalog No. 50976306
Encompass_Preferred
1000mL vacuum trap with #15 Ace-Thred includes round bottom flask, adjustable inner tube, and nylon bushing with FETFE o-ring. Flask dimensions: 125mm x 315mm.
Catalog No. 50-976-306 Supplier Ace Glass, Inc. Supplier No. 874413
1000mL vacuum trap with #15 Ace-Thred includes round bottom flask, adjustable inner tube, and nylon bushing with FETFE o-ring. Flask dimensions: 125mm x 315mm.