Learn More
Ace Glass, Inc. 1000mL vessel with a 48mm O.D. neck to fit a #50 Ace-Thred
SDP
Catalog No. 50973326
Encompass_Preferred
1000mL vessel with a 48mm O.D. neck to fit a #50 Ace-Thred, for connection to an umbrella manifold. Vessel has sloped sides. Working capacity of vessel is about 1/2 of volume.
Catalog No. 50-973-326 Supplier Ace Glass, Inc. Supplier No. 669642
1000mL vessel with a 48mm O.D. neck to fit a #50 Ace-Thred, for connection to an umbrella manifold. Vessel has sloped sides. Working capacity of vessel is about 1/2 of volume.