Learn More
Microsolv Technology Corporation 1000Pk Caps Yellow
Catalog No. NC1509441
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Yellow Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 1000/CS.
Catalog No. NC1509441 Supplier Microsolv Technology Corporation Supplier No. 9502SE35YM
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Yellow Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 1000/CS.