Learn More
Inorganic Ventures 1000µg/mL ALUMINUM HCL 30mL
SDP
Catalog No. NC1667763
Encompass_Preferred
Aluminum/HCl | 1000ug/mL | 2% v/v Hydrochloric Acid | MW: 26.982 | 7429-90-5 | MFCD00134029 | 30mL
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1667763 Supplier Inorganic Ventures Supplier No. CGALCL130ML
Aluminum/HCl | 1000ug/mL | 2% v/v Hydrochloric Acid | MW: 26.982 | 7429-90-5 | MFCD00134029 | 30mL
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search