Learn More
SPEX CERTIPREP LLC 3P Bromide from Sodium Bromide at 1,000 ug/mL in H2O, 100 mL
Catalog No. NC1734465
Encompass_Preferred
(HPS by Zeptometrix) Bromide Anion IC Single-Component Standard from Sodium Bromide at 1,000 ug/mL in H2O.
Catalog No. NC1734465 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICBRM100
(HPS by Zeptometrix) Bromide Anion IC Single-Component Standard from Sodium Bromide at 1,000 ug/mL in H2O.