Learn More
SPEX CERTIPREP LLC 3P 1000PPM PALLADIUM 5% HCI 125ML
Catalog No. NC0802985
Encompass_Preferred
(HPS by Zeptometrix) Pd 1000ug/mL in 5% HCl Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0802985 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000382100
(HPS by Zeptometrix) Pd 1000ug/mL in 5% HCl Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.