Learn More
Inorganic Ventures 1000µg/mL ALUMINUM 30mL
SDP
Catalog No. NC1445762
Encompass_Preferred
1000ug/mL Aluminum | MW: 26.98 | 7429-90-5 | MFCD00134029 | 30mL
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1445762 Supplier Inorganic Ventures Supplier No. CGAL130ML
Add to Cart
Add to Cart
1000ug/mL Aluminum | MW: 26.98 | 7429-90-5 | MFCD00134029 | 30mL
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search