Learn More
Inorganic Ventures 1000µg/mL ANTIMONY 30mL
SDP
Catalog No. NC1445760
Encompass_Preferred
1000 ppm Single Element Antimony Standard for ICP-OES and ICP-MS applications
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1445760 Supplier Inorganic Ventures Supplier No. CGSB130ML
1000 ppm Single Element Antimony Standard for ICP-OES and ICP-MS applications
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search