Learn More
Inorganic Ventures 1000µg/mL CADMIUM 30mL
SDP
Catalog No. NC1492142
Encompass_Preferred
1000 ppm Single Element Cadmium Standard for ICP-OES and ICP-MS applications
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1492142 Supplier Inorganic Ventures Supplier No. CGCD130ML
Add to Cart
Add to Cart
1000 ppm Single Element Cadmium Standard for ICP-OES and ICP-MS applications
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search