Learn More
Inorganic Ventures 1000µg/mL CALCIUM 30mL
SDP
Supplier:  Inorganic Ventures CGCA130ML
Encompass_Preferred
1000 ppm Single Element Calcium Standard for ICP-OES and ICP-MS applications
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1492143