Learn More
SPEX CERTIPREP LLC 3P 1000UG/ML CHLORIDE STD 100ML
Catalog No. NC0634110
Encompass_Preferred
(HPS by Zeptometrix) Chloride IC Std 1000 ug/mL in H2O Exp Date: 18 Months
Catalog No. NC0634110 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICCLM100
(HPS by Zeptometrix) Chloride IC Std 1000 ug/mL in H2O Exp Date: 18 Months