Learn More
Inorganic Ventures 1000µg/mL INDIUM 30mL
SDP
Catalog No. NC1885972
Encompass_Preferred
Matrix: 2% v/v Nitric Acid. Starting Material: In Metal. Analyte: In 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1885972 Supplier Inorganic Ventures Supplier No. CGIN130ML
Matrix: 2% v/v Nitric Acid. Starting Material: In Metal. Analyte: In 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search