Learn More
Alkali Scientific 1000uL Beckman Biomek FX/NX Mu
SDP
Catalog No. 503029427
Encompass_Preferred
Alkali Scientificdesigns its P1000 automation tips to ensure seamless compatibility with Beckman Coulter Biomek FX/NX/iSeries (i5 and i7) and NGeniuSP robotic liquid handlers These tips require no separate labware definition and existing automation control software protocols remain unchanged The Alkali ScientificP1000 tip (1025 L with filter) is a popular choice for Beckman Biomek and NGeniuS liquid handlers It delivers excellent sub-microliter performance (precision and accuracy) while offering a larger volume capacity and length for added protocol flexibility It is compatible with all NGeniuS 96-format heads Alkali Scientificmanufactures its P1000 tips from high-quality virgin polypropylene and uses advanced polishing techniques to minimize liquid retention without the use of releasing agents or additives Tip material Conductive polypropylene Tip box material Polypropylene Compare to Beckman Coulter B85961 B85965 B85959 987936 987935 987925 C59585 Specifications Working volume
Catalog No. 50-302-9427 Supplier Alkali Scientific Supplier No. FXBS1000CS
Alkali Scientificdesigns its P1000 automation tips to ensure seamless compatibility with Beckman Coulter Biomek FX/NX/iSeries (i5 and i7) and NGeniuSP robotic liquid handlers These tips require no separate labware definition and existing automation control software protocols remain unchanged The Alkali ScientificP1000 tip (1025 L with filter) is a popular choice for Beckman Biomek and NGeniuS liquid handlers It delivers excellent sub-microliter performance (precision and accuracy) while offering a larger volume capacity and length for added protocol flexibility It is compatible with all NGeniuS 96-format heads Alkali Scientificmanufactures its P1000 tips from high-quality virgin polypropylene and uses advanced polishing techniques to minimize liquid retention without the use of releasing agents or additives Tip material Conductive polypropylene Tip box material Polypropylene Compare to Beckman Coulter B85961 B85965 B85959 987936 987935 987925 C59585 Specifications Working volume