Learn More
Alkali Scientific 1000uL Beckman Biomek FX/NX Mu
SDP
Catalog No. 503029428
Encompass_Preferred
Alkali Scientificdesigns its P1000 automation tips for optimal compatibility with Beckman Coulter Biomek FX/NX/iSeries (i5 and i7) and NGeniuSP robotic liquid handlers These automation tips do not require a separate labware definition and existing automation control software protocols remain unchanged The Alkali ScientificP1000 tip (1025 L with filter) is widely compatible with Beckman Biomek and NGeniuS liquid handler lines It provides exceptional sub-microliter performance (precision and accuracy) while offering a larger volume capacity and length for enhanced protocol flexibility It is designed to fit all NGeniuS 96-compatible heads Alkali Scientificmanufactures its P1000 tips from premium virgin polypropylene and employs advanced polishing techniques to ensure minimal liquid retention without the use of releasing agents or other additives Tip material Clear polypropylene Tip box material Polypropylene Compare to Beckman Coulter B85961 B85965 B85959 987936 987935 987925 C5958
Catalog No. 50-302-9428 Supplier Alkali Scientific Supplier No. FXF1000CS
Alkali Scientificdesigns its P1000 automation tips for optimal compatibility with Beckman Coulter Biomek FX/NX/iSeries (i5 and i7) and NGeniuSP robotic liquid handlers These automation tips do not require a separate labware definition and existing automation control software protocols remain unchanged The Alkali ScientificP1000 tip (1025 L with filter) is widely compatible with Beckman Biomek and NGeniuS liquid handler lines It provides exceptional sub-microliter performance (precision and accuracy) while offering a larger volume capacity and length for enhanced protocol flexibility It is designed to fit all NGeniuS 96-compatible heads Alkali Scientificmanufactures its P1000 tips from premium virgin polypropylene and employs advanced polishing techniques to ensure minimal liquid retention without the use of releasing agents or other additives Tip material Clear polypropylene Tip box material Polypropylene Compare to Beckman Coulter B85961 B85965 B85959 987936 987935 987925 C5958