Learn More
AUTOMATION TRAINER 1000L Hamilton CO-RE-Type 96-Format, Clear, Filtered, Sterile, Hanging Rack
SDP
Catalog No. NC2219015
Encompass_Preferred
Automation Trainers pipette tips are designed to be a direct replacement to the Hamilton CO-RE Series tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate lab-ware definition is not required.- 50 L, 300 L & 1000 L Max. Working Volumes - High-Grade Polypropylene Construction - 50 L (Case of 60 96-Tip Racks), 300 L (Case of 60 96-Tip Racks), and 1000 L (Case of 40 96-Tip Racks) - Packaging: Case of 12 5-Hanging-Rack Blister Packs (50 L, 300 L), Case of 8 5-Hanging-Rack Blister Packs (1000 L)
This item is not returnable. View return policy
Catalog No. NC2219015 Supplier AUTOMATION TRAINER Supplier No. HC1000CLFSH
Automation Trainers pipette tips are designed to be a direct replacement to the Hamilton CO-RE Series tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate lab-ware definition is not required.- 50 L, 300 L & 1000 L Max. Working Volumes - High-Grade Polypropylene Construction - 50 L (Case of 60 96-Tip Racks), 300 L (Case of 60 96-Tip Racks), and 1000 L (Case of 40 96-Tip Racks) - Packaging: Case of 12 5-Hanging-Rack Blister Packs (50 L, 300 L), Case of 8 5-Hanging-Rack Blister Packs (1000 L)