Learn More
CELLTREAT Scientific Products 1000L Extended Length Low Retention Pipette Tips, Racked, Sterile
SDP
Catalog No. 501656723
Encompass_Preferred
Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Pipette Tips are universal fit and compatible with major pipettes. Smooth, hydrophobic surfaces to ensure accurate pipetting. Extended length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. Molded volume reference markings offer a quick double check opportunity. Hinged lids on rack are designed for easy one-handed opening. Lids have stacking feature for added stability and a secure locking tab. Lightweight lid design prevents tipping when open. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed sterile, 96 tips/rack, 960/case.
Catalog No. 50-165-6723 Supplier CELLTREAT Scientific Products Supplier No. 229037
Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Pipette Tips are universal fit and compatible with major pipettes. Smooth, hydrophobic surfaces to ensure accurate pipetting. Extended length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. Molded volume reference markings offer a quick double check opportunity. Hinged lids on rack are designed for easy one-handed opening. Lids have stacking feature for added stability and a secure locking tab. Lightweight lid design prevents tipping when open. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed sterile, 96 tips/rack, 960/case.