Learn More
CELLTREAT Scientific Products 1000L Extended Length Pipette Tip Rack, Empty Rack for Bulk Tips, Wafer Included, Non-sterile
SDP
Catalog No. 501656740
Encompass_Preferred
Hinged lids on rack are designed for easy one-handed opening, making it easier to access repeatedly. Lids have stacking feature for added stability and a secure locking tab. Lightweight lid design prevents tipping when open. Interior support ribs prevent racks from warping. Empty rack with wafer allows refill from bulk packed pipette tips. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed non-sterile, 20/case.
This item is not returnable. View return policy
Catalog No. 50-165-6740 Supplier CELLTREAT Scientific Products Supplier No. 229068
Hinged lids on rack are designed for easy one-handed opening, making it easier to access repeatedly. Lids have stacking feature for added stability and a secure locking tab. Lightweight lid design prevents tipping when open. Interior support ribs prevent racks from warping. Empty rack with wafer allows refill from bulk packed pipette tips. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed non-sterile, 20/case.