Learn More
Bellco Glass, Inc. 1000 Exten Lgth Low Retentio
SDP
Catalog No. 502329711
Encompass_Preferred
CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Multiple sizes are available including extended length tips 10 and 100 to allow better access to the bottom of test tubes reagent bottles flasks and other vessels.Specifications Pipette Tips are universal fit and compatible with major pipettes Pipette Tips have smooth hydrophobic surfaces to ensure accurate pipetting Molded volume reference markings offer a quick double check opportunity Case Quantity 960
Catalog No. 50-232-9711 Supplier Bellco Glass, Inc. Supplier No. 229022000
CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Multiple sizes are available including extended length tips 10 and 100 to allow better access to the bottom of test tubes reagent bottles flasks and other vessels.Specifications Pipette Tips are universal fit and compatible with major pipettes Pipette Tips have smooth hydrophobic surfaces to ensure accurate pipetting Molded volume reference markings offer a quick double check opportunity Case Quantity 960