Learn More
Bellco Glass, Inc. 1000 Extended Length
SDP
Catalog No. 502330470
Encompass_Preferred
Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly. Reduce plastic waste by refilling empty racks available without a wafer to allow refill from pipette tip reload systems.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20
Catalog No. 50-233-0470 Supplier Bellco Glass, Inc. Supplier No. 229063000
Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly. Reduce plastic waste by refilling empty racks available without a wafer to allow refill from pipette tip reload systems.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20