Learn More
Alkali Scientific 1000uL Hamilton STAR/STARlet N
SDP
Catalog No. 503028744
Encompass_Preferred
Alkali Scientificautomatic transparent 1000 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation Clear CO-RE style tips are made from high-quality polypropylene (PP) polymer with a surface treated to enhance hydrophobicity These transparent tips are meticulously designed for specialized applications delivering improved efficiency and repeatability in your workflows Material Manufactured from medical-grade high-tenacity polypropylene (PP) imported and selected to meet stringent industry standards ensuring material consistency between batches Production Environment Produced in 100000-class cleanroom facilities to prevent contamination ensuring that no DNA RNA protease or endotoxins are present Each batch is traceable with product point collection and packaging minimizing variation and enhancing the accuracy of experimental results Surface Technology Advanced manufacturing processes ensure a smooth inner surface significantly reducing liquid retention fo
Catalog No. 50-302-8744 Supplier Alkali Scientific Supplier No. HST1000
Alkali Scientificautomatic transparent 1000 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation Clear CO-RE style tips are made from high-quality polypropylene (PP) polymer with a surface treated to enhance hydrophobicity These transparent tips are meticulously designed for specialized applications delivering improved efficiency and repeatability in your workflows Material Manufactured from medical-grade high-tenacity polypropylene (PP) imported and selected to meet stringent industry standards ensuring material consistency between batches Production Environment Produced in 100000-class cleanroom facilities to prevent contamination ensuring that no DNA RNA protease or endotoxins are present Each batch is traceable with product point collection and packaging minimizing variation and enhancing the accuracy of experimental results Surface Technology Advanced manufacturing processes ensure a smooth inner surface significantly reducing liquid retention fo