Learn More
Alkali Scientific 1000uL Hamilton STAR/STARlet N
SDP
Catalog No. 503027411
Encompass_Preferred
Alkali Scientificautomatic black conductive 1000 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are crafted from high-quality polypropylene (PP) material with a specially treated surface that enhances hydrophobicity These tips are engineered for precise applications designed to improve efficiency and ensure repeatability in your workflows Material Manufactured from medical-grade high-tenacity polypropylene (PP) sourced and selected according to strict industry standards to ensure consistent material quality between batches Production Process Produced in a 100000-class cleanroom environment to prevent contamination ensuring the absence of DNA RNA protease and endotoxins Every product batch is tracked for traceability with packaging meticulously numbered to ensure consistency and improve experimental accuracy Surface Technology Advanced processing techniques are used to provide a smooth inner surface minimizing liquid retention fo
Catalog No. 50-302-7411 Supplier Alkali Scientific Supplier No. HBFT1000
Alkali Scientificautomatic black conductive 1000 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are crafted from high-quality polypropylene (PP) material with a specially treated surface that enhances hydrophobicity These tips are engineered for precise applications designed to improve efficiency and ensure repeatability in your workflows Material Manufactured from medical-grade high-tenacity polypropylene (PP) sourced and selected according to strict industry standards to ensure consistent material quality between batches Production Process Produced in a 100000-class cleanroom environment to prevent contamination ensuring the absence of DNA RNA protease and endotoxins Every product batch is tracked for traceability with packaging meticulously numbered to ensure consistency and improve experimental accuracy Surface Technology Advanced processing techniques are used to provide a smooth inner surface minimizing liquid retention fo