Learn More
CELLTREAT Scientific Products 1000L Low Retention Pipette Tip Reload System, Non-sterile
SDP
Catalog No. 501656732
Encompass_Preferred
Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Molded volume reference markings offer a quick double check opportunity. Reload system offers clear window for easy identification of color-coded wafers. Support wafers have large grasping tabs for easy removal from Reload System and Rack. Secure locking tab keeps reload system closed and prevents tumbling tips. Reduce plastic waste by refilling empty racks. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed sterile, 960 tips/reload system, 1920/case.
Catalog No. 50-165-6732 Supplier CELLTREAT Scientific Products Supplier No. 229056
Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Molded volume reference markings offer a quick double check opportunity. Reload system offers clear window for easy identification of color-coded wafers. Support wafers have large grasping tabs for easy removal from Reload System and Rack. Secure locking tab keeps reload system closed and prevents tumbling tips. Reduce plastic waste by refilling empty racks. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed sterile, 960 tips/reload system, 1920/case.