Learn More
Research Products International Corp MoleTek 1000ul Universal Non-Filtered Pipet, Polypropylene, 96/RK, 960/case
SDP
Catalog No. 502229556
Encompass
Universal tips are developed to offer a reliable liquid transfer solution for molecular laboratories. High performance tips allow sample recovery and accurate transfer of solution. Molded from high quality medical grade polypropylene (PP) material, manufactured and packed in a clean room (ISO Class 5). Dimensions: - Tip measures 3-1/8” long
Sterile
Graduated
RNase, Dnase Free
CE Marked
Catalog No. 50-222-9556 Supplier Research Products International Corp Supplier No. 147217A
Universal tips are developed to offer a reliable liquid transfer solution for molecular laboratories. High performance tips allow sample recovery and accurate transfer of solution. Molded from high quality medical grade polypropylene (PP) material, manufactured and packed in a clean room (ISO Class 5). Dimensions: - Tip measures 3-1/8” long
Sterile
Graduated
RNase, Dnase Free
CE Marked