Learn More
Genesee Scientific Corporation Olympus Plastics 26-430 1000 and 2000 mL Graduated Cylinders, Pack of 12
SDP
Catalog No. NC1962416
Encompass
The Olympus Plastics 26-430 1000 and 2000 Series Plastic Test Tubes are made of clear polypropylene plastic. They have a round bottom and a screw cap closure. The 1000 series tubes have an outside diameter of 26mm and a height of 43mm. The 2000 series tubes have an outside diameter of 26mm and a height of 75mm. Both tube sizes have a working volume between 15 and 18 mL. The tubes can withstand temperatures from -20 to 121 degrees Celsius. They are sold in cases of 500 tubes. The tubes are suitable for sample storage and transport.
Catalog No. NC1962416 Supplier Genesee Scientific Corporation Supplier No. 26430
The Olympus Plastics 26-430 1000 and 2000 Series Plastic Test Tubes are made of clear polypropylene plastic. They have a round bottom and a screw cap closure. The 1000 series tubes have an outside diameter of 26mm and a height of 43mm. The 2000 series tubes have an outside diameter of 26mm and a height of 75mm. Both tube sizes have a working volume between 15 and 18 mL. The tubes can withstand temperatures from -20 to 121 degrees Celsius. They are sold in cases of 500 tubes. The tubes are suitable for sample storage and transport.