Learn More
Alkali Scientific 1000uL PerkinElmer Revvity Jan
SDP
Catalog No. 503029035
Encompass_Preferred
Alkali Scientifichigh-quality pipette tips are compatible with the Fontus and the JANUS G3 VariSpanpipetting module and can be used for a variety of automated liquid handling applications These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality Tips are certified RNase-/DNase-free and nonpyrogenic Available in nonsterile sterile non-filtered filtered and wide-bore Also available in Maxymum Recovery to further improve accuracy and precision while reducing sample loss Black or Clear polypropylene tips are available in 96-well and 384-well format designed for PerkinElmer Janus/Evolution P /Minitrak/PlateTrak MDT-96 P235 MDT-96 P50 and MDT-384 30 L head The PerkinElmer P1000 conductive black tip supports precise volume transfers up to 1000 L delivering exceptional accuracy and flexibility for various protocols Its design allows access to both 96-format and 384-format plates and is
Catalog No. 50-302-9035 Supplier Alkali Scientific Supplier No. PEBFT1000
Alkali Scientifichigh-quality pipette tips are compatible with the Fontus and the JANUS G3 VariSpanpipetting module and can be used for a variety of automated liquid handling applications These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality Tips are certified RNase-/DNase-free and nonpyrogenic Available in nonsterile sterile non-filtered filtered and wide-bore Also available in Maxymum Recovery to further improve accuracy and precision while reducing sample loss Black or Clear polypropylene tips are available in 96-well and 384-well format designed for PerkinElmer Janus/Evolution P /Minitrak/PlateTrak MDT-96 P235 MDT-96 P50 and MDT-384 30 L head The PerkinElmer P1000 conductive black tip supports precise volume transfers up to 1000 L delivering exceptional accuracy and flexibility for various protocols Its design allows access to both 96-format and 384-format plates and is