Learn More
Alkali Scientific 1000uL PerkinElmer Revvity Jan
SDP
Catalog No. 503028351
Encompass_Preferred
Alkali Scientificdesigns its PerkinElmer automation tips for seamless compatibility with PerkinElmer Janus Varispan liquid handlers Using our PerkinElmer Janus-compatible automation tips eliminates the need for a separate labware definition and there is no requirement to modify PerkinElmer Janus automation control software protocols This conductive black 96-format tip is designed to be fully interchangeable with the original PerkinElmer P1000 automation pipette tip Additionally it is compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThe PerkinElmer P1000 conductive black tip is designed for volume transfers up to 1000 L and provides outstanding precision and accuracy Its capacity and length offer flexibility in protocol design allowing it to access both 96-format and 384-format plates These tips are compatible with Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P1000 tips f
Catalog No. 50-302-8351 Supplier Alkali Scientific Supplier No. PEFT1000
Alkali Scientificdesigns its PerkinElmer automation tips for seamless compatibility with PerkinElmer Janus Varispan liquid handlers Using our PerkinElmer Janus-compatible automation tips eliminates the need for a separate labware definition and there is no requirement to modify PerkinElmer Janus automation control software protocols This conductive black 96-format tip is designed to be fully interchangeable with the original PerkinElmer P1000 automation pipette tip Additionally it is compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThe PerkinElmer P1000 conductive black tip is designed for volume transfers up to 1000 L and provides outstanding precision and accuracy Its capacity and length offer flexibility in protocol design allowing it to access both 96-format and 384-format plates These tips are compatible with Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P1000 tips f