Learn More
CELLTREAT Scientific Products 1000L Pipette Tips, LTS Fit, Sterile
SDP
Catalog No. NC2206448
Encompass_Preferred
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tips. Pipette tips are packaged in racks with hinged lids designed for easy one-handed opening. 10μL tips feature an extended length for easier access to the bottom of vessels. Fine tip for improved accuracy and precision. Filter tips are fitted with a polymer barrier to reduce the risk of contamination and prevent aerosols from penetrating into the pipette. Lids feature stacking supports, and are removable to save bench space.
This item is not returnable. View return policy
Catalog No. NC2206448 Supplier CELLTREAT Scientific Products Supplier No. 229076
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tips. Pipette tips are packaged in racks with hinged lids designed for easy one-handed opening. 10μL tips feature an extended length for easier access to the bottom of vessels. Fine tip for improved accuracy and precision. Filter tips are fitted with a polymer barrier to reduce the risk of contamination and prevent aerosols from penetrating into the pipette. Lids feature stacking supports, and are removable to save bench space.