Learn More
Genesee Scientific Corporation 1000UL REACH
SDP
Catalog No. NC1832111
Encompass
1000μl Reach Classic Filter Pipette TipsLow Binding, Racked, Sterile40 Racks of 96 Pipet Tips/Unit
Catalog No. NC1832111 Supplier Genesee Scientific Corporation Supplier No. 24430C
1000μl Reach Classic Filter Pipette TipsLow Binding, Racked, Sterile40 Racks of 96 Pipet Tips/Unit