Learn More
Waters Corp Waters Sample Loop, 1 mL, HPLC
Catalog No. NC1762221 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 1 mL Sample Loop is a large volume sample loop and is standard with the Prep Option Kit.
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Type: Loop
  • System type: HPLC
  • Volume: 1 mL
  • Replacement schedule: As required
Catalog No. NC1762221 Supplier Waters Corp Supplier No. 700002462
This 1 mL Sample Loop is a large volume sample loop and is standard with the Prep Option Kit.
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Type: Loop
  • System type: HPLC
  • Volume: 1 mL
  • Replacement schedule: As required