Learn More
Genesee Scientific Corporation 1000µl Olympus Ergo X Barrier Tip for Rainin™ LTS™ | 10 Racks of 96 Tips/Unit
SDP
Catalog No. NC1718250
Encompass
Olympus Plastics Ergonomic X Pipet Tips are specifically designed and engineered for use with the Rainin™ LTS™ LiteTouch™ Pipettes and are precision manufactured to the industry’s most rigorous quality control standards. All tips are made from virgin polypropylene and contain no recycled, reprocessed, or altered plastics that might introduce contamination or defects. Every lot of each product can be traced to the corresponding raw material and production date. All Olympus Tips are INDEPENDENTLY CERTIFIED RNase, DNase, Human DNA, PCR Inhibitor and Pyrogen-FREE. In addition to the in-process control monitoring, all product lots are sampled after packaging and tested in our QC laboratory AND independent laboratories, thus resulting in a contaminant-free product of the highest quality.
Catalog No. NC1718250 Supplier Genesee Scientific Corporation Supplier No. 24729
Olympus Plastics Ergonomic X Pipet Tips are specifically designed and engineered for use with the Rainin™ LTS™ LiteTouch™ Pipettes and are precision manufactured to the industry’s most rigorous quality control standards. All tips are made from virgin polypropylene and contain no recycled, reprocessed, or altered plastics that might introduce contamination or defects. Every lot of each product can be traced to the corresponding raw material and production date. All Olympus Tips are INDEPENDENTLY CERTIFIED RNase, DNase, Human DNA, PCR Inhibitor and Pyrogen-FREE. In addition to the in-process control monitoring, all product lots are sampled after packaging and tested in our QC laboratory AND independent laboratories, thus resulting in a contaminant-free product of the highest quality.