Learn More
Genesee Scientific Corporation Genesee Scientific 1000µl XL Tips, Low Binding, Sterile, Universal Fit, 8 Racks of 96 Tips
SDP
Catalog No. NC2400191
Encompass
Genesee Scientific 1000µl XL Premium Barrier Tips are high-quality, sterile, low-binding pipette tips designed for accurate and contaminant-free liquid handling in scientific research. Their proprietary low-binding polymer technology minimizes sample adhesion while preventing pipettor contamination, ensuring reliable performance in sensitive applications. Compatible with most research-grade pipettors, these tips offer enhanced accuracy and are certified free of RNase, DNase, and DNA.
Catalog No. NC2400191 Supplier Genesee Scientific Corporation Supplier No. 23430
Genesee Scientific 1000µl XL Premium Barrier Tips are high-quality, sterile, low-binding pipette tips designed for accurate and contaminant-free liquid handling in scientific research. Their proprietary low-binding polymer technology minimizes sample adhesion while preventing pipettor contamination, ensuring reliable performance in sensitive applications. Compatible with most research-grade pipettors, these tips offer enhanced accuracy and are certified free of RNase, DNase, and DNA.