Learn More
Teknova 1000X Trace Metals, 250mL, Sterile. 8 Pack.
SDP
Catalog No. 502745844
Encompass
A trace metals solution is a mixture that contains a small concentration of metal ions. These solutions have many uses—from hardening water to removing impurities from metals, and purifying semiconductors. Trace metals solutions are used to culture cells and grow bacteria. By controlling the amount of metal ions in the solution, scientists can create an environment that is ideal for cell growth.
Catalog No. 50-274-5844 Supplier Teknova Supplier No. 213368PK
A trace metals solution is a mixture that contains a small concentration of metal ions. These solutions have many uses—from hardening water to removing impurities from metals, and purifying semiconductors. Trace metals solutions are used to culture cells and grow bacteria. By controlling the amount of metal ions in the solution, scientists can create an environment that is ideal for cell growth.