Learn More
Sino Biological Recombinant Anti-SOX2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100106R017100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the human Sox2.
Catalog No. 50-161-0449