Learn More
Sino Biological Recombinant Human Neuropilin-1/NRP1 Protein (ECD, V179A, hFc Tag)
Supplier:  Sino Biological 10011H02H50
Encompass_Preferred
A DNA sequence encoding the human NRP1 isoform b (NP_001019799.1, with natural variant Val 179 Ala) (Met1-Lys644) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3781