Learn More
Sino Biological Anti-Neuropilin-1/NRP1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10011MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Neuropilin-1 / NRP1 / CD304 (rh Neuropilin-1 / NRP1 / CD304; Catalog#10011-H08H; NP_001019799.1; Met1-Lys644). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0461