Learn More
Sino Biological Recombinant Human VEGFR2/KDR Protein (Domain 2&3, hFc Tag)
Supplier:  Sino Biological 10012H02H2200
Encompass_Preferred
A DNA sequence encoding the human KDR (NP_002244.1) (Asp120-Lys327) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3787