Learn More
Sino Biological Recombinant Human VEGFR2/KDR Protein (Domain 1&2&3, His Tag)
Supplier:  Sino Biological 10012H08H1200
Encompass_Preferred
A DNA sequence encoding the human KDR (NP_002244.1) (Met1-Lys327) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3791