Learn More
Sino Biological Anti-VEGFR2/KDR Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10012MM03F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VEGFR2/KDR/Flk-1/CD309 (rh VEGFR2/KDR/Flk-1/CD309; Catalog#10012-H08H; NP_002244.1; Met1-Glu764) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-0518