Learn More
Sino Biological Recombinant Anti-VEGFR2/KDR Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10012R001P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGFR2/KDR/Flk-1/CD309 (rh VEGFR2/KDR/Flk-1/CD309; Catalog#10012-H08H; NP_002244.1; Met1-Glu764) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-0539