Learn More
Sino Biological Recombinant Anti-Bcl-2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100126R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, the synthetic peptide corresponding to the N-terminus of the Human Bcl-2.
Catalog No. 50-161-0482