Learn More
Sino Biological Recombinant Anti-Bcl-2 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 100126R204F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human Bcl-2, and it was conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-0491