Learn More
Sino Biological Recombinant Anti-Bcl-2 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 100126R204P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human BCL2/Bcl-2, and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-0493