Learn More
Sino Biological Anti-Bcl-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100126T36100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the Human Bcl-2. Human Bcl-2 specific IgG was purified by Human Bcl-2 affinity chromatography.
Catalog No. 50-161-0494