Learn More
Sino Biological Recombinant Anti-CD79A Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100128R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the C-terminus of the human CD79a.
Catalog No. 50-161-0497